Learn More
Cytiva Amersham™ Hybond™ Membranes
Unsupported 100% pure nitrocellulose membrane validated for use with all ECL Western and protein blotting applications
Marque: Cytiva RPN303D
Code : GE
Informations supplémentaires : CAS Number : 9004-70-0 Weight : 0.65000kg
Description
- Excellent sensitivity, resolution and low background for all labeling and detection systems: radioactive, nonradioactive and chromogenic
- Packaging features include interleaf on both sides of the membrane and airtight resealable bag with marking spot
- Pore size: 0.2μm and 0.45μm
- Discs are also available
Spécification
30 cm | |
30cm x 3m | |
1 Roll | |
Nitrocellulose |
3 m | |
Protein blotting | |
Roll | |
0.45 μm |