missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Hybond™ Membranes

Unsupported 100% pure nitrocellulose membrane validated for use with all ECL Western and protein blotting applications

Brand:  Cytiva RPN82D

216.60 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.



 View more versions of this product

Product Code. 11758398

  • £235.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Excellent sensitivity, resolution and low background for all labeling and detection systems: radioactive, nonradioactive and chromogenic
  • Packaging features include interleaf on both sides of the membrane and airtight resealable bag with marking spot
  • Pore size: 0.2μm and 0.45μm
  • Discs are also available

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8.2cm
82 mm
Disc
0.45 μm
Protein blotting
50 Discs
Nitrocellulose
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.