missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Hybond™ PVDF Membranes: Sheets
Ideal for Western blotting applications where stripping and reprobing are necessary
£82.40 - £407.00
Product Code | Brand | Dimensions (LxW) | Format | Quantity | Description | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | Format | Quantity | Description | Price | Quantity & Availability | ||
15279834
|
Cytiva
10600057 |
20 x 20 cm | Sheet | 25/Pk. | Hybond P |
£407.00
25 sheets |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
15289834
|
Cytiva
10600058 |
20 x 20 cm | Sheet | 10/Pk. | Hybond P |
£251.00
10 sheets |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
15219874
|
Cytiva
10600101 |
8 x 9 cm | Sheet | 25/Pk. | Hybond P |
£121.61
25 sheets |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
15299884
|
Cytiva
10600060 |
20 x 20 cm | Sheet | 10/Pk. | Hybond LFP |
£289.00
10 sheets |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
15229894
|
Cytiva
10600102 |
8 x 9 cm | Sheet | 25/Pk. | Hybond LFP |
£162.00
25 sheets |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||
15229864
|
Cytiva
10600089 |
10 x 10 cm | Sheet | 10/Pk. | Hybond P |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15229824
|
Cytiva
10600039 |
26 x 60 cm | Sheet | 5/Pk. | Hybond P |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15289884
|
Cytiva
10600040 |
26.5 x 60 cm | Sheets (LFP) | 5/Pk. | Hybond LFP |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15299834
|
Cytiva
10600061 |
20 x 20 cm | Sheets (Seq) | 10/Pk. | Hybond SEQ |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15219864
|
Cytiva
10600088 |
10 x 10 cm | Sheet | 25/Pk. | Hybond P |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Individual sheets.
- Hydrophobic PVDF membranes
- Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights
Amersham Hybond 0.2μm
- Compatible with a range of solvents used for rapid destaining
- Specifically designed for protein sequencing applications and solid phase amino acid analysis
- Offers excellent blotting for proteins Mr<20,000
- Specifically optimized for fluorescence detection in Western blotting applications
- Provides low background fluorescence, resulting in high sensitivity
Specifications
PVDF |