missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls

Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Brand:  Cytiva 10600001

281.67 GBP valid until 2025-12-31
Use promo code "24363" to get your promotional price.



 View more versions of this product

Product Code. 15249794

  • £342.00 / 1 roll

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.

Packed in rolls.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

TRUSTED_SUSTAINABILITY
Specifications

Specifications

4 m
Western blots, dot-blot assays, and other protein or nucleic acid methods
100% Pure Nitrocellulose
4 m x 30 cm
30 cm
Roll
0.2 μm
Amersham Protran
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : Nitrocellulose

SDS Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls > 0.2um; Roll; 30cm x 4m

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.