Learn More
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Brand: Cytiva 10600002
217.20 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : GH
Additional Details : CAS Number : 9004-70-0 Weight : 0.06600kg
Description
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Packed in rolls.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
4 m | |
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
100% Pure Nitrocellulose | |
4 m x 30 cm |
30 cm | |
Roll | |
0.45 μm | |
Amersham Protran |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : Nitrocellulose