missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls
Shop All Cytiva Products
Click to view available options
Width (Metric):
15 cm
20 cm
30 cm
Pore Size:
0.1 μm
0.2 μm
0.45 μm
Unit Size:
1 roll
Description
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Packed in rolls.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
Specifications
| Length (Metric) | 4 m |
| Width (Metric) | 20 cm |
| For Use With (Application) | Western blots, dot-blot assays, and other protein or nucleic acid methods |
| Format | Roll |
| Material | 100% Pure Nitrocellulose |
| Pore Size | 0.1 μm |
Safety and Handling
Product Identifier
- Amersham Protran
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction