Learn More
Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Brand: Cytiva 10600093
96.43 GBP valid until 2025-12-31
Use promo code "24363" to get your promotional price.
Description
Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.
Individual sheets.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
8 cm | |
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
Sheet |
9 cm | |
25/Pk. | |
0.45 μm |
Safety and Handling
- Amersham Protran 0.45
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.