missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Brand:  Cytiva 10600125

99.29 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.



 View more versions of this product

Product Code. 15265928

  • £108.00 / 10 sheets

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10.2 cm
Western blots, dot-blot assays, and other protein or nucleic acid methods
Sheet
0.2 μm
13.3 cm
10/Pk.
100% Pure Nitrocellulose
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers