missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Brand:  Cytiva 10600044

195.70 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.



Code : GH

Additional Details : CAS Number : 9004-70-0 Weight : 0.02200kg


 View more versions of this product

Product Code. 15269824

  • £206.00 / 10 sheets
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

Use for Western blots, dot-blot assays, and other protein or nucleic acid methods. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability
  • Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent

Specifications

Specifications

20 cm
Western blots, dot-blot assays, and other protein or nucleic acid methods
Sheet
20 cm
10/Pk.
0.2 μm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers