missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ NC Nitrocellulose Western Blotting Sandwiches

Convenient, precut, preassembled Protran NC Nitrocellulose membrane and filter paper sandwiches

Brand:  Cytiva 10600115

120.65 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.



 View more versions of this product

Product Code. 15209884

  • £130.68 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
  • Fit most precase gel formats

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8 cm
Pack of 10
0.2 μm
9 cm
Sandwich
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger||

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.