Learn More
Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls
Use for the best results with fluorescence Western blotting detection applications. Cytiva Life Sciences™ Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Brand: Cytiva 10600008
192.60 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : GH
Additional Details : CAS Number : 9004-70-0 Weight : 0.04400kg
Description
Supplied in rolls.
- Low background enables a dynamic range for complicated samples
- Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
Specifications
4 m | |
Western blots, dot-blot assays, and other protein or nucleic acid methods | |
100% Pure Nitrocellulose |
20 cm | |
Roll | |
0.45 μm |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose