missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets
Provides exceptional handling characteristics, allowing reprobing
Brand: Cytiva 10600051
This item is not returnable.
View return policy
Description
Individual sheets.
- Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
- The support does not affect transfer conditions or results
Specifications
20 cm | |
25/Pk. | |
Nitrocellulose, Inert Polyester |
20 cm | |
Sheet | |
0.45 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- amersham protran supported 0.45µm NC 200mm x 200mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose