Learn More
Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets
Provides exceptional handling characteristics, allowing reprobing
Brand:  Cytiva 10600054
 View more versions of this product
Product Code. 15269834
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Individual sheets.
  • Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
  • The support does not affect transfer conditions or results
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 cm
10/Pk.
Nitrocellulose, Inert Polyester
20 cm
Sheet
0.2 μm