Learn More
Merck Amicon™ Ultra-0.5 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
Brand: Merck UFC501096
459.98 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 72
Additional Details : Weight : 0.36000kg
Includes: Two microcentrifuge tubes
Description
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
- High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
- Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
- Heat-sealed membrane eliminates possibility of downstream extractables
- Universal rotor compatibility
- High retentate recovery of >90%
- High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
- Easy sample monitoring due to translucent housing and volume gradations
- Direct pipetter sample access eliminates processing step to recover concentrate
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates
- Concentration of dilute or prepurified proteins from column eluents
- Desalting and buffer exchange
Specifications
Single-use | |
10 kDa | |
1 cm2 | |
Regenerated Cellulose (RC) | |
14,000 xG for concentration spin; 1000 xG for reverse spin | |
Centrifugal Filters | |
0.196 in. | |
10.8 mm |
High-recovery Ultracel-PL Regenerated Cellulose | |
Non-sterile | |
Protein purification, protein concentration and desalting | |
Two microcentrifuge tubes | |
96 Pack | |
0.5 mL | |
4.99 cm |