missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Ultra-0.5 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
£64.80 - £3090.00
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | High-recovery Ultracel-PL Regenerated Cellulose |
Sterility | Non-sterile |
Filtration Area | 1 cm2 |
For Use With (Application) | Protein purification, protein concentration and desalting |
Product Code | Brand | MWCO | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
10795785
|
Merck
UFC500308 |
3 kDa | 0.5 mL | 8Pack |
£124.00
Pack of 8 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||
10212584
|
Merck
UFC500324 |
3 kDa | 0.5 mL | 24Pack |
£290.00
Pack of 24 |
In Stock
Log in to see stock. |
|||
10704645
|
Merck
UFC500396 |
3 kDa | 0.5 mL | 96Pack |
£1010.00
Pack of 96 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||
10130884
|
Merck
UFC5003BK |
3 kDa | 0.5 mL | 500Pack |
£3090.00
Pack of 500 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
10028713
|
Merck
UFC501008 |
10 kDa | 0.5 mL | 8Pack |
£68.60
Pack of 8 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||
10088753
|
Merck
UFC501024 |
10 kDa | 0.5 mL | 24Pack |
£294.12
Pack of 24 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||
10282244
|
Merck
UFC501096 |
10 kDa | 0.5 mL | 96 Pack |
£1040.00
Pack of 96 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||
10150644
|
Merck
UFC5010BK |
10 kDa | 0.5 mL | 500Pack |
£2198.16
Pack of 500 |
N/A | |||
Currently Unavailable: We have no information when this product will be available
Alternative Produkte anzeigen
|
10505274
|
Merck
UFC503008 |
30 kDa | 0.5 mL | 8Pack |
£69.15
Pack of 8 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
10012584
|
Merck
UFC503024 |
30 kDa | 0.5 mL | 24Pack |
£282.00
Pack of 24 |
In Stock
Log in to see stock. |
|||
10256744
|
Merck
UFC503096 |
30 kDa | 0.5 mL | 96Pack |
£916.00
Pack of 96 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
10328054
|
Merck
UFC5030BK |
30 kDa | 0.5 mL | 500Pack |
£2245.00
Pack of 500 |
N/A | |||
Currently Unavailable: We have no information when this product will be available
Alternative Produkte anzeigen
|
10378434
|
Merck
UFC505008 |
50 kDa | 0.5 mL | 8Pack |
£64.80
Pack of 8 |
N/A | ||
Currently Unavailable: We have no information when this product will be available
Alternative Produkte anzeigen
|
10194304
|
Merck
UFC505024 |
50 kDa | 0.5 mL | 24Pack |
£294.12
Pack of 24 |
In Stock
Log in to see stock. |
||
10695445
|
Merck
UFC505096 |
50 kDa | 0.5 mL | 96Pack |
£880.00
Pack of 96 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||
10257324
|
Merck
UFC5050BK |
50 kDa | 0.5 mL | 500Pack |
£2255.00
Pack of 500 |
Estimated Shipment: 25-10-2024
Log in to see stock. |
|||
10527164
|
Merck
UFC510008 |
100 kDa | 0.5 mL | 8Pack |
£111.00
Pack of 8 |
N/A | |||
Currently Unavailable: We have no information when this product will be available
Alternative Produkte anzeigen
|
10114694
|
Merck
UFC510024 |
100 kDa | 0.5 mL | 24Pack |
£277.61
Pack of 24 |
In Stock
Log in to see stock. |
||
10388244
|
Merck
UFC510096 |
100 kDa | 0.5 mL | 96Pack |
£1035.00
Pack of 96 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
10398244
|
Merck
UFC5100BK |
100 kDa | 0.5 mL | 500Pack |
£2120.00
Pack of 500 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
Beschreibung
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
- High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
- Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
- Heat-sealed membrane eliminates possibility of downstream extractables
- Universal rotor compatibility
- High retentate recovery of >90%
- High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
- Easy sample monitoring due to translucent housing and volume gradations
- Direct pipetter sample access eliminates processing step to recover concentrate
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates
- Concentration of dilute or prepurified proteins from column eluents
- Desalting and buffer exchange
Spezifikation
Single-use | |
Non-sterile | |
Protein purification, protein concentration and desalting | |
Two microcentrifuge tubes | |
Centrifugal Filters | |
0.196 in. | |
10.8 mm |
High-recovery Ultracel-PL Regenerated Cellulose | |
1 cm2 | |
Regenerated Cellulose (RC) | |
14,000 xG for concentration spin; 1000 xG for reverse spin | |
0.5 mL | |
4.99 cm |