Learn More
Thermo Scientific™ AminoLink™ Coupling Resin
Covalently immobilize proteins and antibodies by lysine residues or other primary amines (-NH2) via reductive amination with these aldehyde-activated agarose beads.
£160.00 - £441.00
Specifications
Description | AminoLink Coupling Resin |
---|---|
Content And Storage | Store at 4°C. |
Product Line | AminoLink™ |
Target | Antibody, Proteins through Primary Amine |
Type | Resin |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11826774
|
Thermo Scientific™
20381 |
AminoLink Coupling Resin | 10 mL |
£160.00
10mL |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
11836774
|
Thermo Scientific™
20382 |
AminoLink Coupling Resin | 50 mL |
£441.00
50mL |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
Description
Thermo Scientific™ AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups to enable covalent immobilization of antibodies and other proteins through primary amines.
AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups. Proteins and other molecules with primary amines can be covalently attached (immobilized) to the resin to make chromatography columns for use in affinity purification. The aldehyde groups form stable secondary amine bonds with primary amines such as exist in the side chain of lysine (K) residues, which are generally abundant and readily accessible in proteins. Once a protein is immobilized, the prepared affinity resin can be used for a variety of batch and column affinity purification methods involving binding interactions with the immobilized protein. The resin and linkage are stable in most binding and elution conditions typically used in affinity chromatography, enabling prepared resin to be used for multiple rounds of affinity purification procedures.Highlights:
AminoLink Coupling Resin – aldehyde-activated crosslinked 4% beaded agarose
Ideal for antibodies and other proteins – immobilize molecules via primary amines (-NH2)
Flexible coupling conditions – efficient (>85%) coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer with or without organic solvent
Stable, permanent immobilization – coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand
Better than immobilization to CNBr-activated agarose – bond is more stable and uncharged, resulting in less nonspecific binding in affinity purification procedures
Versatile and reusable – prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions
Includes:
Kit contains pre-packed AminoLink Columns, coupling buffer, quenching buffer, wash solution, sodium cyanoborohydride solution, and column accessories
Specifications
AminoLink Coupling Resin | |
AminoLink™ | |
Resin | |
Bottle |
Store at 4°C. | |
Antibody, Proteins through Primary Amine | |
Aldehyde | |
Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.