missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Amintra (R) Desalting Spin Columns

£559.00 - £929.00

Specifications

Content And Storage 4°C
For Use With (Application) Flow Cytometry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18061634
View Documents
Novus Biologicals™
ADS0050
50 Units
£559.00
50 units
Please to purchase this item. Need a web account? Register with us today!
18063944
View Documents
Novus Biologicals™
ADS0100
100 Units
£929.00
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

4°C
Flow Cytometry
Videos
SDS
Documents

Documents

Product Certifications