Learn More
Ammonia, ca. 7N solution in methanol, Thermo Scientific Chemicals
Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger
Brand: Thermo Scientific Acros 133710251
1480.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Ammonia | |
ca. 7N Solution in Methanol | |
H3N | |
6.0 to 7.5N | |
05,15 | |
15, 488 | |
QGZKDVFQNNGYKY-UHFFFAOYSA-N | |
N | |
17.03 | |
222 | |
17.03 | |
0.77 |
Colorless to Yellow | |
7664-41-7, 67-56-1 | |
NH3 | |
MFCD00011418 | |
0.7700g/mL | |
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol | |
25 L | |
azane | |
14°C | |
CHEBI:16134 | |
Plastic Can | |
Liquid |
Safety and Handling
- Ammonia, ca 7N solution in methanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2411519 | Certificate of Analysis | 11/01/2025 | 133710025,133710010,133710250,133710251 |
BS24/028 | Certificate of Analysis | 23/12/2024 | 133710025,133710010,133710250,133710251 |
2421799 | Certificate of Analysis | 23/12/2024 | 133710025,133710010,133710250,133710251 |
2372583 | Certificate of Analysis | 14/03/2024 | 133710025,133710010,133710250,133710251 |
2365468 | Certificate of Analysis | 14/03/2024 | 133710025,133710010,133710250,133710251 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only