missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Aluminum Sulfate Dodecahydrate, Reagent Grade, ≥99% (Titration), Honeywell™
Reagent Grade,≥99% (titration)
£130.06 - £210.50
Chemical Identifiers
CAS | 7784-26-1 |
---|---|
Molecular Formula | AlH28NO20S2 |
Molecular Weight (g/mol) | 453.313 |
MDL Number | MFCD00149958 |
InChI Key | WZUKKIPWIPZMAS-UHFFFAOYSA-K |
Synonym | ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate |
PubChem CID | 62668 |
IUPAC Name | aluminum;azanium;disulfate;dodecahydrate |
SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15607640
|
Honeywell Research Chemicals
A2140-500G |
500 g | Poly Bottle |
£130.06
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15697630
|
Honeywell Research Chemicals
A2140-1KG |
1 kg | Poly Bottle |
£210.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7784-26-1 | |
453.313 | |
WZUKKIPWIPZMAS-UHFFFAOYSA-K | |
62668 | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3] |
AlH28NO20S2 | |
MFCD00149958 | |
ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate | |
aluminum;azanium;disulfate;dodecahydrate |
Specifications
93.5°C | |
White | |
AlH28NO20S2 | |
MFCD00149958 | |
ammonia alum, unii-5c36drl9zn, aluminum ammonium sulfate dodecahydrate, aluminum ammonium disulfate dodecahydrate, ammonium aluminum sulfate hydrate, alum, ammonium usp, aluminum ammonium sulfate dodecahydrate, ammonium aluminum sulfate dodecahydrate, 5c36drl9zn, ammonium aluminum disulfate dodecahydrate | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3] | |
453.313 | |
453.33g/mol | |
Odorless | |
1.640g/cm3 at 20°C |
7784-26-1 | |
2.6 (at 25°C) | |
AlNH4(SO4)2 · 12H2O | |
NONH for all modes of transport | |
WZUKKIPWIPZMAS-UHFFFAOYSA-K | |
aluminum;azanium;disulfate;dodecahydrate | |
62668 | |
≥99% (titration) | |
Crystalline | |
Ammonium aluminum sulfate dodecahydrate |
Safety and Handling
missing translation for 'rtecsNumber' : 232-055-3