missing translation for 'onlineSavingsMsg'
Learn More

Ammonium benzoate, 99%, Thermo Scientific Chemicals

£22.91 - £139.70

Chemical Identifiers

CAS 1863-63-4
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00013069
InChI Key VWSRWGFGAAKTQG-UHFFFAOYSA-N
Synonym ammonium benzoate, benzoic acid, ammonium salt, ammonii benzoas, ammonium benzoicum, vulnoc ab, benzoic acid ammonium salt, hsdb 555, unii-ac80wd7gpf, benzoic acid, ammonium salt 1:1, ac80wd7gpf
PubChem CID 15830
IUPAC Name azanium;benzoate
SMILES C1=CC=C(C=C1)C(=O)[O-].[NH4+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11327348
SDS View Documents
Thermo Scientific Alfa Aesar
030753.22
100 g
£22.91
100g
Please to purchase this item. Need a web account? Register with us today!
11307348
SDS View Documents
Thermo Scientific Alfa Aesar
030753.36
500 g
£45.63
500g
Please to purchase this item. Need a web account? Register with us today!
11317348
SDS View Documents
Thermo Scientific Alfa Aesar
030753.A3
2 kg
£139.70
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ammonium benzoate is used predominantly in industries which acts as a corrosion inhibitors and anti-scaling agents . It is also used as paint additives and coating additives. Other consumer uses include cleaning and furnishing Care products, paints and coatings, personal care products, water treatment products. Ammonium benzoate is also used as chemical analysis reagents, in electronics industry, used for the production of electrolytic capacitors and also used in pharmaceutical industries. Used as adhesives, metal rust treatment and rubber additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 217-468-9

missing translation for 'rtecsNumber' : DG3378000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium benzoate, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.