missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Cerium(Iv) Sulfate Dihydrate, ≥98.0% (RT), puriss. p.a. ACS Reagent, Honeywell Fluka™ Product Code.: 15682190

Ammonium Cerium(Iv) Sulfate Dihydrate, ≥98.0% (RT), puriss. p.a. ACS Reagent, Honeywell Fluka™

Product Code. 15682190 Shop All Honeywell Fluka Products
100 g, Poly Bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15682190

Brand: Honeywell-Fluka 22269100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥98.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-47-9
MDL Number MFCD00149417

Specifications

Quantity 100 g
Linear Formula Ce(NH4)4(SO4)4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 632.55g/mol
Percent Purity ≥98.0% (RT)
Grade ACS/puriss. p.a.
Packaging Poly Bottle
Chemical Name or Material Ammonium Cerium(Iv) Sulfate Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Cerium(Iv) Sulfate Dihydrate, ≥98.0% (RT), puriss. p.a. ACS Reagent, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.