missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, 99.6%, ACS reagent, Thermo Scientific Chemicals

£30.85 - £182.91

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10444505
View Documents
Thermo Scientific Acros
423285000
500 g Plastic bottle
£30.85
500g
Please to purchase this item. Need a web account? Register with us today!
10404711
View Documents
Thermo Scientific Acros
423280010
1 kg Plastic bottle
£48.64
1kg
Please to purchase this item. Need a web account? Register with us today!
10063773
View Documents Promotion Details
Thermo Scientific Acros
423280050
5 kg Plastic Drum
£182.91
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
N.Cl
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206

Specifications

340.0°C
White
Crystalline Powder
ClH4N
MFCD00011420
15, 506
Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate
N.Cl
53.49
CHEBI:31206
99.6%
0.01% max
Ammonium chloride
12125-02-9
4.5 to 5.5 (5% soln. at 25°C)
99.6%
NH3·HCl
15,13
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
NLXLAEXVIDQMFP-UHFFFAOYSA-N
azanium;chloride
25517
53.49
ACS Reagent
1.5200g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not eat,d

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 235-186-4

Product Identifier
  • Ammonium chloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.