missing translation for 'onlineSavingsMsg'
Learn More
Ammonium chloride, 99.6%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10404711

Ammonium chloride, 99.6%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10404711
1 kg, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle

Product Code. 10404711

Brand: Thermo Scientific Acros 423280010

Please to purchase this item. Need a web account? Register with us today!

Edge

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumkloridShow MoreShow Less
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340.0°C
Color White
pH 4.5 to 5.5 (5% soln. at 25°C)
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99.6%
Linear Formula NH3·HCl
Fieser 15,13
Merck Index 15, 506
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, practically insoluble in acetone, ether and, ethyl acetate
IUPAC Name amine hydrochloride
Formula Weight 53.49
Percent Purity 99.6%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.01% max
Density 1.5200g/mL
Chemical Name or Material Ammonium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460680 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number423280010,423285000,423280050
Lot NumberA0457336 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number423280010,423285000,423280050
Lot NumberA0396560 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number
Lot NumberA0410304 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number423280010,423285000,423280050
Lot NumberA0446030 Certificate TypeCertificate of Analysis Date28/09/2022 Catalog Number
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 24126

    Expires2025-02-13

    Days

    0

    Hours

    21

    Minutes

    23

    Seconds

    11

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium chloride, 99.6%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.