Learn More
Ammonium dichromate, 99%, Thermo Scientific Chemicals
£20.60 - £152.76
Chemical Identifiers
CAS | 7789-09-5 |
---|---|
Molecular Formula | H8Cr2N2O7 |
Molecular Weight (g/mol) | 252.07 |
MDL Number | MFCD00010879 |
InChI Key | JOSWYUNQBRPBDN-UHFFFAOYSA-P |
Synonym | ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 |
PubChem CID | 24600 |
IUPAC Name | diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
SMILES | [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10752295
|
Thermo Scientific Acros
208811000 |
100 g | Plastic bottle |
£20.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10276260
|
Thermo Scientific Acros
208815000 |
500 g | Plastic bottle |
£45.02
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10126683
|
Thermo Scientific Acros
208810025 |
2.5 kg | Plastic bottle |
£152.76
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7789-09-5 | |
252.07 | |
JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
24600 | |
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
H8Cr2N2O7 | |
MFCD00010879 | |
ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 | |
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |
Specifications
170.0°C | |
Orange | |
98.5% min. (Iodometry) | |
(NH4)2Cr2O7 | |
15, 512 | |
Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol | |
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] | |
252.07 | |
252.07 | |
3% max. (105°C, 4 hrs) |
7789-09-5 | |
Adhering Crystals | |
H8Cr2N2O7 | |
MFCD00010879 | |
ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481 | |
JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24600 | |
99% | |
Ammonium dichromate, 99% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
May damage fertility.
May damage the unborn child.
May cause genetic defects.
Causes damage to organs through prolonge
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-143-1
missing translation for 'rtecsNumber' : HX7650000
missing translation for 'tsca' : TSCA
RUO – Research Use Only