missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 99+%, for analysis, Thermo Scientific Chemicals

£29.04 - £615.06

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10742051
View Documents
Thermo Scientific Acros
201832500
250 g Plastic bottle
£29.04
250g
Please to purchase this item. Need a web account? Register with us today!
10082910
View Documents
Thermo Scientific Acros
201830010
1 kg Plastic bottle
£78.29
1kg
Please to purchase this item. Need a web account? Register with us today!
10063953
View Documents
Thermo Scientific Acros
201830025
2.5 kg Plastic bottle
£176.88
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15199213
View Documents
Thermo Scientific Acros
201830100
10 kg Plastic Drum
£615.06
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7722-76-1
115.03
LFVGISIMTYGQHF-UHFFFAOYSA-N
24402
phosphoric acid amine
H6NO4P
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
CHEBI:62982
N.OP(O)(O)=O

Specifications

190.0°C
White
Crystalline Solid
H6NO4P
MFCD00003396
ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
LFVGISIMTYGQHF-UHFFFAOYSA-N
phosphoric acid amine
24402
115.03
Analytical
Ammonium dihydrogen phosphate
7722-76-1
4 to 4.5 (5% soln. at 20°C)
99+%
NH4H2PO4
15,541
Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
N.OP(O)(O)=O
115.03
CHEBI:62982
≥99%
1.8g/mL
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-764-5

Product Identifier
  • Ammonium dihydrogen phosphate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only