missing translation for 'onlineSavingsMsg'
Learn More
Ammonium iron(III) citrate, brown, Thermo Scientific Chemicals Product Code.: 11463504

Ammonium iron(III) citrate, brown, Thermo Scientific Chemicals

Product Code. 11463504
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11463504

Brand: Thermo Scientific Alfa Aesar A11199.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.

Solubility
Soluble in water.1200 g/L (20°C)

Notes
Hygroscopic. Desiccate at room temperature. Store in the dark. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7+3
Molecular Weight (g/mol) 264.999
MDL Number MFCD00013099
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltzShow MoreShow Less
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

Color Brown
Quantity 250 g
Merck Index 14,4017
Solubility Information Soluble in water.1200g/L (20°C)
Odor Weak
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Ammonium iron (III) citrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466392 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog NumberA11199.0I,A11199.0B,A11199.30
Lot NumberA0451297 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog NumberA11199.0I,A11199.0B,A11199.30
Lot NumberU17J023 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog NumberA11199.0I,A11199.0B,A11199.30
Lot NumberH8786A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11199.0I,A11199.0B,A11199.30
Lot NumberU21F048 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11199.0I,A11199.0B,A11199.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium iron(III) citrate, brown, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.