Learn More
Ammonium iron(III) sulfate dodecahydrate, 99+%, for analysis, Thermo Scientific Chemicals
£18.59 - £126.63
Chemical Identifiers
CAS | 7783-83-7 |
---|---|
Molecular Formula | H4FeNO8S2·12H2O |
Molecular Weight (g/mol) | 482.19 |
MDL Number | MFCD00150004 |
InChI Key | LCPUDZUWZDSKMX-UHFFFAOYSA-K |
Synonym | ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g |
PubChem CID | 197097 |
IUPAC Name | azanium;iron(3+);disulfate;dodecahydrate |
SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10331054
|
Thermo Scientific Acros
205880500 |
50 g | Plastic bottle |
£18.59
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10791224
|
Thermo Scientific Acros
205881000 |
100 g | Plastic bottle |
£18.89
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10124740
|
Thermo Scientific Acros
205885000 |
500 g | Plastic bottle |
£38.99
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10584274
|
Thermo Scientific Acros
205880025 |
2.5 kg | Plastic bottle |
£126.63
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-83-7 | |
482.19 | |
LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
197097 | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
H4FeNO8S2·12H2O | |
MFCD00150004 | |
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
azanium;iron(3+);disulfate;dodecahydrate |
Specifications
230.0°C | |
7783-83-7 | |
Adhering Crystalline Powder or Crystals | |
H4FeNO8S2·12H2O | |
MFCD00150004 | |
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
azanium;iron(3+);disulfate;dodecahydrate | |
197097 | |
≥99% | |
1.71 | |
Ammonium iron(III) sulfate dodecahydrate, p.a. |
39.0°C to 41.0°C | |
Gray-Purple | |
99% min. (Iodometry) | |
(NH4)Fe(SO4)2·12H2O | |
15, 515 | |
Solubility in water: 1240g/L water (25°C). Other solubilities: practically insoluble in alcohol | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
482.19 | |
482.19 | |
Analytical | |
1.7100g/mL |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
missing translation for 'rtecsNumber' : WS5900000
- Ammonium iron(III) sulfate dodecahydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Iron(III) ammonium sulfate dodecahydrate
RUO – Research Use Only