Learn More
Ammonium magnesium phosphate hydrate, 99.999% (metals basis), Thermo Scientific Chemicals
£103.00 - £408.00
Chemical Identifiers
CAS | 7785-21-9 |
---|---|
Molecular Formula | H4MgNO4P |
Molecular Weight (g/mol) | 137.31 |
MDL Number | MFCD00050358 |
InChI Key | MXZRMHIULZDAKC-UHFFFAOYSA-L |
Synonym | unii-76yp0j930v, magnesium ammonium phosphate, ammonium magnesium phosphate hydrate, ammonium magnesium orthophosphate, phosphoric acid, ammonium magnesium salt, azanium magnesium phosphate, phosphoric acid, ammonium magnesium salt 1:1:1, magnesium 2+ ammonium phosphate, magnesium 2+ ion ammonium phosphate, ammonium magnesium phosphate nh4 mg po4 |
PubChem CID | 178727 |
SMILES | [NH4+].[Mg++].[O-]P([O-])([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11355118
|
Thermo Scientific Alfa Aesar
044053.18 |
50 g |
£103.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365118
|
Thermo Scientific Alfa Aesar
044053.30 |
250 g |
£408.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ammonium magnesium phosphate hydrate general application in pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7785-21-9 | |
137.31 | |
MXZRMHIULZDAKC-UHFFFAOYSA-L | |
178727 |
H4MgNO4P | |
MFCD00050358 | |
unii-76yp0j930v, magnesium ammonium phosphate, ammonium magnesium phosphate hydrate, ammonium magnesium orthophosphate, phosphoric acid, ammonium magnesium salt, azanium magnesium phosphate, phosphoric acid, ammonium magnesium salt 1:1:1, magnesium 2+ ammonium phosphate, magnesium 2+ ion ammonium phosphate, ammonium magnesium phosphate nh4 mg po4 | |
[NH4+].[Mg++].[O-]P([O-])([O-])=O |
Specifications
7785-21-9 | |
H4MgNO4P | |
unii-76yp0j930v, magnesium ammonium phosphate, ammonium magnesium phosphate hydrate, ammonium magnesium orthophosphate, phosphoric acid, ammonium magnesium salt, azanium magnesium phosphate, phosphoric acid, ammonium magnesium salt 1:1:1, magnesium 2+ ammonium phosphate, magnesium 2+ ion ammonium phosphate, ammonium magnesium phosphate nh4 mg po4 | |
MXZRMHIULZDAKC-UHFFFAOYSA-L | |
magnesium(2+) ammonium phosphate | |
178727 | |
99.999% | |
(metals basis) |
Powder | |
MFCD00050358 | |
Soluble in water. | |
[NH4+].[Mg++].[O-]P([O-])([O-])=O | |
137.31 | |
137.32 (Anhydrous) | |
1.711 g/mL | |
Ammonium magnesium phosphate hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 232-075-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only