missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium molybdate, 99.99%, (trace metal basis), Thermo Scientific Chemicals
£72.00 - £720.00
Chemical Identifiers
CAS | 13106-76-8 |
---|---|
Molecular Formula | H8MoN2O4 |
Molecular Weight (g/mol) | 196.01 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12357113
|
Thermo Scientific Acros
448540050 |
5 g | Glass bottle |
£72.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12367113
|
Thermo Scientific Acros
448540250 |
25 g | Glass bottle |
£246.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12377113
|
Thermo Scientific Acros
448541000 |
100 g | Glass bottle |
£720.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13106-76-8 | |
H8MoN2O4 | |
196.01 |
Specifications
13106-76-8 | |
H8MoN2O4 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
196.01 | |
Trace Metal Basis |
99.99% | |
(NH4)2MoO4 | |
196.01 | |
99.99% | |
Ammonium molybdate |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 236-031-3
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ammonium molybdate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only