missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Nitrate, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥98%
£36.98 - £115.00
Chemical Identifiers
CAS | 6484-52-2 |
---|---|
Molecular Formula | H4N2O3 |
Molecular Weight (g/mol) | 80.04 |
MDL Number | MFCD00011425 |
InChI Key | DVARTQFDIMZBAA-UHFFFAOYSA-O |
Synonym | ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills |
PubChem CID | 22985 |
ChEBI | CHEBI:63038 |
IUPAC Name | azanium;nitrate |
SMILES | [NH4+].[O-][N+]([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
6484-52-2 | |
80.04 | |
DVARTQFDIMZBAA-UHFFFAOYSA-O | |
22985 | |
azanium;nitrate |
H4N2O3 | |
MFCD00011425 | |
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills | |
CHEBI:63038 | |
[NH4+].[O-][N+]([O-])=O |
Specifications
6484-52-2 | |
NH4NO3 | |
1942 | |
DVARTQFDIMZBAA-UHFFFAOYSA-O | |
azanium;nitrate | |
22985 | |
80.04g/mol | |
ACS/puriss. p.a. | |
≤0.01% (as SO4) |
H4N2O3 | |
MFCD00011425 | |
ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills | |
[NH4+].[O-][N+]([O-])=O | |
80.04 | |
CHEBI:63038 | |
≥98% | |
Plastic Bottle | |
Ammonium Nitrate, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P220-P261-P305 + P351 + P338
H272-H315-H319-H335
missing translation for 'einecsNumber' : 229-347-8
missing translation for 'rtecsNumber' : BR9050000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ammonium nitrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title