Learn More
Ammonium persulfate, 98+%, Thermo Scientific Chemicals
Oxidation reagent
£24.02 - £680.39
Chemical Identifiers
CAS | 7727-54-0 |
---|---|
Molecular Formula | H8N2O8S2 |
Molecular Weight (g/mol) | 228.19 |
MDL Number | MFCD00003390 |
InChI Key | ROOXNKNUYICQNP-UHFFFAOYSA-N |
Synonym | ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french |
PubChem CID | 62648 |
IUPAC Name | diazanium;sulfonatooxy sulfate |
SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10341681
|
Thermo Scientific Acros
401160100 |
10 g | Glass Bottle |
£24.02
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10219790
|
Thermo Scientific Acros
401161000 |
100 g | Plastic bottle |
£29.15
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10095750
|
Thermo Scientific Acros
401165000 |
500 g | Plastic bottle |
£32.46
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10483731
|
Thermo Scientific Acros
401160020 |
2 kg | Glass bottle |
£86.23
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10651705
|
Thermo Scientific Acros
401160050 |
5 kg | Plastic Bottle |
£184.92
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10712164
|
Thermo Scientific Acros
401160250 |
25 kg | Fibre drum |
£680.39
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Oxidation reagentChemical Identifiers
7727-54-0 | |
228.19 | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
62648 | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
H8N2O8S2 | |
MFCD00003390 | |
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
diazanium;sulfonatooxy sulfate |
Specifications
120.0°C | |
White | |
98+% | |
(NH4)2S2O8 | |
01,952; 02,348; 03,238; 05,15; 06,20; 12,33 | |
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
228.19 | |
228.19 | |
ACS Reagent | |
1.98 | |
>120°C | |
Ammonium persulfate |
7727-54-0 | |
Crystalline Powder and/or Crystals | |
H8N2O8S2 | |
MFCD00003390 | |
15, 539 | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
diazanium;sulfonatooxy sulfate | |
62648 | |
≥98% | |
0.05% max. | |
Negligible | |
1.9800g/mL |
Safety and Handling
GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if swallowed.
May intensify fire; oxidizer.
GHS P Statement:
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Ammonium persulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only