missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, purum p.a., Honeywell™
Purum p.a.,≥99.0% (T)
£16.43 - £109.61
Chemical Identifiers
CAS | 7783-20-2 |
---|---|
Molecular Formula | H8N2O4S |
Molecular Weight (g/mol) | 132.13 |
MDL Number | MFCD00003391 |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
PubChem CID | 6097028 |
ChEBI | CHEBI:62946 |
IUPAC Name | diazanium;sulfate |
SMILES | N.N.OS(O)(=O)=O |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
6097028 | |
diazanium;sulfate |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
CHEBI:62946 | |
N.N.OS(O)(=O)=O |
Specifications
7783-20-2 | |
(NH4)2SO4 | |
NONH for all modes of transport | |
Soluble in water, 0.5/10g/mL | |
N.N.OS(O)(=O)=O | |
132.13 | |
CHEBI:62946 | |
≥99% (T) | |
1.77g/mL at 25°C |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
diazanium;sulfate | |
6097028 | |
132.14g/mol | |
purum p.a. | |
Ammonium Sulfate |
Safety and Handling
missing translation for 'einecsNumber' : 231-984-1
missing translation for 'rtecsNumber' : BS4500000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title