Learn More
Ammonium tetrafluoroborate, 99%, pure, Thermo Scientific Chemicals
£18.60 - £192.00
Chemical Identifiers
CAS | 13826-83-0 |
---|---|
Molecular Formula | BF4H4N |
Molecular Weight (g/mol) | 104.84 |
MDL Number | MFCD06245785 |
InChI Key | PDTKOBRZPAIMRD-UHFFFAOYSA-O |
Synonym | ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p |
PubChem CID | 9964072 |
SMILES | [NH4+].F[B-](F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10369902
|
Thermo Scientific Acros
310961000 |
100 g | Plastic bottle |
£18.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10393003
|
Thermo Scientific Acros
310965000 |
500 g | Plastic bottle |
£48.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13438508
|
Thermo Scientific Acros
310960025 |
2.5 kg | Plastic bottle |
£192.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13826-83-0 | |
104.84 | |
PDTKOBRZPAIMRD-UHFFFAOYSA-O | |
9964072 |
BF4H4N | |
MFCD06245785 | |
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p | |
[NH4+].F[B-](F)(F)F |
Specifications
230.0°C | |
Colorless | |
99% | |
NH4BF4 | |
05,17; 06,132 | |
Solubility in water: soluble. Other solubilities: soluble in ammonium hydride | |
[NH4+].F[B-](F)(F)F | |
104.84 | |
104.84 | |
Pure |
13826-83-0 | |
Crystalline Powder | |
BF4H4N | |
MFCD06245785 | |
ammonium tetrafluoroborate, ammonium fluoroborate, ammonium fluoborate, ammonium borofluoride, unii-c945z80o8x, ammonium tetrafluroborate, azanium tetrafluoroborate, arnmonium tetrafluoroborate, nh4bf4, ksc492c2p | |
PDTKOBRZPAIMRD-UHFFFAOYSA-O | |
azanium;tetrafluoroborate | |
9964072 | |
99% | |
Ammonium tetrafluoroborate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 237-531-4
RUO – Research Use Only