Learn More
Ammonium titanyl oxalate monohydrate, 98%, pure, Thermo Scientific Chemicals
£42.41 - £294.47
Chemical Identifiers
CAS | 10580-03-7 |
---|---|
Molecular Formula | C4H8N2O9Ti |
Molecular Weight (g/mol) | 275.98 |
MDL Number | MFCD00149620 |
InChI Key | VBUNWMBOVJSSMH-UHFFFAOYSA-L |
Synonym | Ammonium bis(oxalato)oxotitanate(IV) |
IUPAC Name | diammonium oxotitaniumbis(ylium) dioxalate |
SMILES | [NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10764733
|
Thermo Scientific Acros
212730100 |
10 g | Glass Bottle |
£42.41
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10140452
|
Thermo Scientific Acros
212731000 |
100 g | Plastic bottle |
£294.47
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10580-03-7 | |
275.98 | |
VBUNWMBOVJSSMH-UHFFFAOYSA-L | |
diammonium oxotitaniumbis(ylium) dioxalate |
C4H8N2O9Ti | |
MFCD00149620 | |
Ammonium bis(oxalato)oxotitanate(IV) | |
[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
Specifications
10580-03-7 | |
Crystalline Powder and/or Crystals | |
C4H8N2O9Ti | |
MFCD00149620 | |
Ammonium bis(oxalato)oxotitanate(IV) | |
VBUNWMBOVJSSMH-UHFFFAOYSA-L | |
diammonium oxotitaniumbis(ylium) dioxalate | |
294.02 | |
Pure | |
Ammonium titanyl oxalate monohydrate |
White to Beige | |
98% | |
(NH4)2TiO(C2O4)2·H2O | |
10,584 | |
Solubility in water: soluble | |
[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O | |
275.98 | |
98% | |
Authentic |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
- Ammonium titanyl oxalate monohydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.