Learn More
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™
For selection for ampicillin resistance in mutated and transformed cells
£49.80 - £141.00
Chemical Identifiers
CAS | 69-52-3 |
---|---|
Molecular Formula | C16H21N3NaO4S |
Molecular Weight (g/mol) | 374.411 |
InChI Key | BSFVNXCYXDYHOD-ZQDFAFASSA-N |
Synonym | Ampicillin |
PubChem CID | 131673879 |
IUPAC Name | (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodiumShow More |
SMILES | [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10193433
|
Fisher BioReagents
BP1760-5 |
5 g | Poly Bottle |
£49.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10419313
|
Fisher BioReagents
BP1760-25 |
25 g | Poly Bottle |
£141.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For selection for ampicillin resistance in mutated and transformed cells
For selection for ampicillin resistance in mutated and transformed cells. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.Chemical Identifiers
69-52-3 | |
374.411 | |
Ampicillin | |
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodiumShow More |
C16H21N3NaO4S | |
BSFVNXCYXDYHOD-ZQDFAFASSA-N | |
131673879 | |
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] |
Specifications
Ampicillin Sodium Salt | |
69-52-3 | |
215°C | |
2% Max. | |
C16H21N3NaO4S | |
Ampicillin | |
[HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na] | |
374.411 | |
371.39 |
Solid | |
8.0 to 10.0 | |
Inclusive between 845μg/mg | |
White | |
15, 583 | |
BSFVNXCYXDYHOD-ZQDFAFASSA-N | |
(2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium | |
131673879 | |
Ampicillin Sodium Salt |
Safety and Handling
WARNING!
Emergency Overview
May cause allergic respiratory and skin reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:2
Flammability:0
Instability:0
Recommended Storage : 4°C
- Ampicillin Sodium Salt
Signal Word
- Danger
Hazard Category
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.