Learn More
OHAUS™ Ampoule Tube Holder
OHAUS™ Ampoule Tube Holder is an accessory for use with vortex mixers.
Brand: OHAUS™ 30400208
Additional Details : Weight : 0.90000kg
Description
- Appplications include: re-suspending liquids, mixing reagents, buffers and emulsions, ELISA assays, drug extractions, mixing samples, PCR, digestions, cell disruptions, genotyping
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the holder for your sample size, or select the flat insert and cut out to suit your custom specifications
Specifications
10 to 17 mm Ampoule Tubes | |
Ampoule Tube Holder | |
Ampoule Tube Holder - Large (10 to 17 mm) | |
Large (10 to 17 mm) |
10 to 17 mm | |
Stainless Steel | |
Vortexer |