missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Ampoule Tube Holder
OHAUS™ Ampoule Tube Holder is an accessory for use with vortex mixers.
£114.00 - £163.00
Specifications
Product Type | Ampoule Tube Holder |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Vortexer |
Product Code | Brand | Size | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Holds | Price | Quantity & Availability | ||||
15792619
|
OHAUS™
30400207 |
Small (15 to 17 mm) | 15 to 17 mm Ampoule Tubes |
£114.00
Each |
Estimated Shipment: 20-11-2024
Log in to see stock. |
||||
15702629
|
OHAUS™
30400208 |
Large (10 to 17 mm) | 10 to 17 mm Ampoule Tubes |
£163.00
Each |
In Stock
Log in to see stock. |
||||
Description
- Appplications include: re-suspending liquids, mixing reagents, buffers and emulsions, ELISA assays, drug extractions, mixing samples, PCR, digestions, cell disruptions, genotyping
- Increase throughput by mixing multiple samples at one time
- Effortlessly mix up to 52 microtubes or eight microplates
- Choose the holder for your sample size, or select the flat insert and cut out to suit your custom specifications
Specifications
Ampoule Tube Holder | |
Stainless Steel | |
Vortexer |