Learn More
Agilent BioTek AMX Automated Media Exchange
GREENER_CHOICE
AMX Automated Media Exchange module protects 3D cell structures (e.g. spheroids, tumoroids), and suspension cells
Brand:  Agilent BioTek 1260041
Product Code. 16592102
  • ÂŁ8140.00 / Each
Explore more special offers
Description
Description
  • AMX Automated Media Exchange module protects 3D cell structures (e.g. spheroids, tumoroids), and suspension cells
  • For use with Agilent BioTek Microplate readers and imagers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE and TUV marked. RoHS compliant.
Primary/Secondary Screening Assays,Compound Storage,Genomics and Proteomics Research,ELISA,Cell Based Washing, Fixing, Staining, Volume or Concentration Normalization
AMX Autmated Media Exchange module, second peri pump, AMX dispense and aspirate manifolds
AMX Automated Media Exhange module
MFXP2, MFXP2R
Product Suggestions
Product Suggestions
Videos
Videos
  • BioTek Automated Media Exchange (AMX) for Spheroid Cellular Models
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      BioTek Automated Media Exchange (AMX) for Spheroid Cellular Models
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Agilent BioTek AMX Automated Media Exchange > use with MFXP2, MFXP2R