Learn More
Interscience™ Anabac Deodorizing Capsules
Placed in an autoclave, Anabac capsules provide a discreet way to perfume the laboratory.
Brand:  Interscience™ 320100
 View more versions of this product
Product Code. 10422084
  • £84.95 / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Capsule dissolves during an autoclaving destruction cycle
  • Capsule melts completely without leaving residue
  • Eliminates unpleasant odors that can emerge from the autoclave
Specifications
Specifications
Autoclave Capsule
Capsules
Cap
Sterile
Autoclavable
Single-use
Refreshes and Perfumes Air Where Autoclave Is Located
100 Capsules/Pk.
Yes
Plastic
Fresh Scent
Yellow
Clear
Non-filtered
Autoclave
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Interscience™ Anabac Deodorizing Capsules > Anabac Deodorizing Capsules; Appearance: Yellow; Color: Clear; Scent: Fresh Scent; Sterility: Sterile