Learn More
BINDER™ Analog Output
Non-adjustable analog output
Brand: BINDER™ 8012-2375
This item is not returnable.
View return policy
Description
- Analog output
- Not adjustable
- For temperature values or temperature and humidity values
- 4 to 20 mA
Specifications
KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, KB PRO 1600 | |
Copper | |
Analog Output | |
For Temperature and Humidity Values |
Product Content Correction
Product Title
BINDER™ Analog Output > For use with KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, KB PRO 1600
Spot an opportunity for improvement?Share a Content Correction