Learn More
Abeba™ Anatom 2125 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 2125/47
Additional Details : Weight : 1.10800kg
 View more versions of this product
Product Code. 15607827
  • £84.95 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 SB A, E SRC
  • Microfiber black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3582)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • Suitable for heel and foot lift
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E SRC
Microfiber
Slip-resistant
2125
Black
47
Polyurethane
Steel