Learn More
Abeba™ Anatom 2135 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 2135/39
This item is not returnable.
View return policy
Description
- CE EN ISO 20345:2011 S2 SRC
- Microfiber black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area adjustable, with elastic band
- Suitable for heel and foot lift
Specifications
CE EN ISO 20345:2011 S2 SRC | |
Microfiber | |
Slip-resistant | |
2135 |
Black | |
39 | |
Polyurethane | |
Steel |