Learn More
Abeba™ Anatom 32247 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32247/37
Additional Details : Weight : 0.95000kg
 View more versions of this product
Product Code. 15699517
  • £98.82 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S1P SRC
  • Velours with breathable textile inlays
  • Dark blue
  • Steel toe cap
  • Shock absorbant PU midsole
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Scratch-resistant coating in toe area
Specifications
Specifications
CE EN ISO 20345:2011 S1P SRC
Velour
Slip-resistant
32247
Blue
37
Polyurethane
Steel