Learn More
ABEBA™ Anatom Black/Red Safety Shoes S3 ESD
Red/black Anatom shoes made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
Brand:  ABEBA™ 32238/45
Additional Details : Weight : 1.28000kg
 View more versions of this product
Product Code. 17101215
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Comfortable, robust and durable shoes
  • Comply to EN ISO 20345: 2012, S3, SRC
  • ESD dissipative according to EN ISO 61340
  • Slip resistant PU double density outsole
  • Black/red water and tear resistant Cordura
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Scratch-resistant coated fabric in toe and heel area
  • Breathable and antibacterial lining with silver ions
  • Removable active comfort insole
Specifications
Specifications
Black, Red
45
EN ISO 20345: 2012 S3 SRC, EN ISO 61340 ESD class 1
Low-cut
S3 SRC ESD
Cordura
Steel
Men's
Water Resistant