Learn More
ABEBA™ Anatom Green/Yellow Safety Shoes S3
Robust, durable, made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture midsole
Brand: ABEBA™ 2251/42
Additional Details : Weight : 1.16000kg
Description
- Comply to EN ISO 20345: 2012, S3, SRC
- Slip resistant PU double density outsole
- Green/yellow water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable active comfort insole
Specifications
Green, Yellow | |
42 | |
EN ISO 20345: 2012 S3 SRC | |
Low-cut | |
S3 SRC |
Cordura | |
Steel | |
Men's | |
Water Resistant |