missing translation for 'onlineSavingsMsg'
Learn More

Aniline-2-sulfonic Acid, 95%, Thermo Scientific Chemicals

£18.90 - £156.00

Chemical Identifiers

CAS 88-21-1
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.19
MDL Number MFCD00007705
InChI Key ZMCHBSMFKQYNKA-UHFFFAOYSA-N
Synonym orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
PubChem CID 6926
ChEBI CHEBI:1015
IUPAC Name 2-aminobenzenesulfonic acid
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10147180
View Documents
Thermo Scientific Acros
400640250
25 g Glass bottle
£18.90
25g
Please to purchase this item. Need a web account? Register with us today!
10246993
View Documents
Thermo Scientific Acros
400641000
100 g Glass bottle
£43.50
100g
Please to purchase this item. Need a web account? Register with us today!
10514054
View Documents
Thermo Scientific Acros
400645000
500 g Glass bottle
£156.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

88-21-1
173.19
ZMCHBSMFKQYNKA-UHFFFAOYSA-N
6926
2-aminobenzenesulfonic acid
C6H7NO3S
MFCD00007705
orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
CHEBI:1015
C1=CC=C(C(=C1)N)S(=O)(=O)O

Specifications

88-21-1
White to Beige
95%
H2NC6H4SO3H
14, 681
orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
ZMCHBSMFKQYNKA-UHFFFAOYSA-N
2-aminobenzenesulfonic acid
6926
173.19
Crystalline Powder
>300°C
Authentic
C6H7NO3S
MFCD00007705
15, 6978
Solubility in water: 3100mg/l (23°C)
C1=CC=C(C(=C1)N)S(=O)(=O)O
173.19
CHEBI:1015
95%
Aniline-2-sulfonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-810-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.