Learn More
Aniline, 99+%, Thermo Scientific Chemicals
Description
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.
Solubility
Miscible with water.
Notes
Light sensitive. Incompatible with oxidizing agents, iron and iron salts.

Specifications
Specifications
Melting Point | -6°C |
Density | 1.021 |
Boiling Point | 183°C to 184°C |
Flash Point | 70°C (158°F) |
Refractive Index | 1.586 |
Quantity | 1000 g |
UN Number | UN1547 |
Beilstein | 605631 |
Merck Index | 14,659 |
Solubility Information | Miscible with water. |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253614 | Certificate of Analysis | 16/01/2025 | A14443.22,A14443.0I,A14443.0B |
10253561 | Certificate of Analysis | 24/12/2024 | A14443.22,A14443.0I,A14443.0B |
10242759 | Certificate of Analysis | 09/08/2022 | A14443.22,A14443.0I,A14443.0B |
10241247 | Certificate of Analysis | 20/06/2022 | A14443.22,A14443.0I,A14443.0B |
10237670 | Certificate of Analysis | 12/04/2022 | A14443.22,A14443.0I,A14443.0B |
Safety and Handling




- Aniline
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Aniline
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.