missing translation for 'onlineSavingsMsg'
Learn More
Anthrone, 95%, Thermo Scientific Chemicals Product Code.: 11439416

Anthrone, 95%, Thermo Scientific Chemicals

Product Code. 11439416
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11439416

Brand: Thermo Scientific Alfa Aesar A19118.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.

Solubility
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-44-8
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00001187
InChI Key RJGDLRCDCYRQOQ-UHFFFAOYSA-N
Synonym anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744Show MoreShow Less
PubChem CID 7018
ChEBI CHEBI:33835
SMILES O=C1C2=CC=CC=C2CC2=CC=CC=C12

Specifications

Melting Point 153°C to 160°C
Quantity 100 g
Beilstein 1910173
Sensitivity Light sensitive
Merck Index 14,691
Solubility Information Soluble in alcohol, benzene, and hot NaOH solutions; Insoluble in water
IUPAC Name 9,10-dihydroanthracen-9-one
Formula Weight 194.23
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Anthrone, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459775 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberA19118.14,A19118.36,A19118.22
Lot NumberA0421301 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberA19118.22
Lot Number10240258 Certificate TypeCertificate of Analysis Date25/04/2022 Catalog NumberA19118.14,A19118.36,A19118.22
Lot Number10197947 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19118.14,A19118.36,A19118.22
Lot Number10156183 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19118.14,A19118.36,A19118.22
compliance-icons
Product Identifier
  • Anthrone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 9(10H)-Anthracenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Anthrone, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.