missing translation for 'onlineSavingsMsg'
Learn More

BD anti-Allophycocyanin (APC) Magnetic Particles - DM IMag-DM, Clone: E30-221

Brand:  BD 557932

Additional Details : Weight : 0.02000kg

Product Code. 15869118

  • £775.00 / Each
Estimated Shipment: 12-09-2024
to see stock.
Add to basket

Description

Description

Cell Separation

Specifications

Specifications

Store undiluted at 4°C.
5 mL
Cell separation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers