missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse anti-Human, V450, Clone: B-LY6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560369

 View more versions of this product

Product Code. 15880139

  • £545.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Reacts with the 150kDa adhesion glycoprotein (p150, integrin α chain) expressed on NK cells, a subset of B and T cells, monocytes, granulocytes and macrophages. It associates with CD18 to form the CD11c/CD18 complex, which binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 may play a role as an adhesion molecule which binds to receptors on stimulated epithelium. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Mouse
Clone: B-ly6
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD11c
Monoclonal
Violet Fluor 450 (V450)
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Spleen Cells from Human with Hairy Cell Leukemia
120 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
B-LY6
ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.